missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| L-Ribose | |
| 90°C | |
| C5H10O5 | |
| 25 g | |
| HMFHBZSHGGEWLO-ZHZWXSSZNA-N | |
| (3S,4R,5S)-5-(hydroxymethyl)oxolane-2,3,4-triol | |
| 90428 | |
| 150.13 | |
| Crystalline Powder |
| 24259-59-4 | |
| White | |
| MFCD00167010 | |
| l-ribose, l-+-ribose, 2s,3s,4s-2,3,4,5-tetrahydroxypentanal, aldehydo-l-ribose, aldehydo-l-ribo-pentose, ror, l-ribose 9ci, ksc202k1p | |
| OC[C@@H]1OC(O)[C@@H](O)[C@H]1O | |
| 150.13 | |
| CHEBI:47015 | |
| ≥98.0% (HPLC) |
Chemical Identifiers
| 24259-59-4 | |
| 150.13 | |
| HMFHBZSHGGEWLO-ZHZWXSSZNA-N | |
| 90428 | |
| (3S,4R,5S)-5-(hydroxymethyl)oxolane-2,3,4-triol |
| C5H10O5 | |
| MFCD00167010 | |
| l-ribose, l-+-ribose, 2s,3s,4s-2,3,4,5-tetrahydroxypentanal, aldehydo-l-ribose, aldehydo-l-ribo-pentose, ror, l-ribose 9ci, ksc202k1p | |
| CHEBI:47015 | |
| OC[C@@H]1OC(O)[C@@H](O)[C@H]1O |
Safety and Handling
EINECSNumber : (5)-6838
TSCA : No