missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Ribose 98.0+%, TCI America™
$32.28 - $227.84
Chemical Identifiers
| CAS | 24259-59-4 |
|---|---|
| Molecular Formula | C5H10O5 |
| Molecular Weight (g/mol) | 150.13 |
| MDL Number | MFCD00167010 |
| InChI Key | HMFHBZSHGGEWLO-ZHZWXSSZNA-N |
| Synonym | l-ribose, l-+-ribose, 2s,3s,4s-2,3,4,5-tetrahydroxypentanal, aldehydo-l-ribose, aldehydo-l-ribo-pentose, ror, l-ribose 9ci, ksc202k1p |
| PubChem CID | 90428 |
| ChEBI | CHEBI:47015 |
| IUPAC Name | (3S,4R,5S)-5-(hydroxymethyl)oxolane-2,3,4-triol |
| SMILES | OC[C@@H]1OC(O)[C@@H](O)[C@H]1O |
Chemical Identifiers
| 24259-59-4 | |
| 150.13 | |
| HMFHBZSHGGEWLO-ZHZWXSSZNA-N | |
| 90428 | |
| (3S,4R,5S)-5-(hydroxymethyl)oxolane-2,3,4-triol |
| C5H10O5 | |
| MFCD00167010 | |
| l-ribose, l-+-ribose, 2s,3s,4s-2,3,4,5-tetrahydroxypentanal, aldehydo-l-ribose, aldehydo-l-ribo-pentose, ror, l-ribose 9ci, ksc202k1p | |
| CHEBI:47015 | |
| OC[C@@H]1OC(O)[C@@H](O)[C@H]1O |
Specifications
| 24259-59-4 | |
| White | |
| MFCD00167010 | |
| l-ribose, l-+-ribose, 2s,3s,4s-2,3,4,5-tetrahydroxypentanal, aldehydo-l-ribose, aldehydo-l-ribo-pentose, ror, l-ribose 9ci, ksc202k1p | |
| OC[C@@H]1OC(O)[C@@H](O)[C@H]1O | |
| 150.13 | |
| CHEBI:47015 | |
| ≥98.0% (HPLC) | |
| L-Ribose |
| 90°C | |
| C5H10O5 | |
| 1 g | |
| HMFHBZSHGGEWLO-ZHZWXSSZNA-N | |
| (3S,4R,5S)-5-(hydroxymethyl)oxolane-2,3,4-triol | |
| 90428 | |
| 150.13 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-6838
TSCA : No