Learn More
Thermo Scientific Chemicals L-(+)-Rhamnose monohydrate, 98+%
CAS: 10030-85-0 | C6H12O5 | 164.16 g/mol
$75.56 - $593.67
Chemical Identifiers
| CAS | 10030-85-0 |
|---|---|
| Molecular Formula | C6H12O5 |
| Molecular Weight (g/mol) | 164.16 |
| MDL Number | MFCD00149363,MFCD00136036 |
| InChI Key | SHZGCJCMOBCMKK-HGVZOGFYSA-N |
| Synonym | l-+-rhamnose monohydrate, 2r,3r,4s,5s-2,3,4,5-tetrahydroxyhexanal hydrate, l + rhamnopyranose, l-mannose, 6-deoxy-, monohydrate, 6-deoxy-l-mannose hydrate, rhamnose hydrate, l-rha hydrate, l-rhamnose hydrate, l +-rhamnose hydrate, a-l-rhamnose monohydrate |
| PubChem CID | 20849066 |
| SMILES | C[C@@H]1O[C@@H](O)[C@H](O)[C@H](O)[C@H]1O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1616606
|
Thermo Scientific Chemicals
A1616606 |
5 g |
Each for $75.56
|
|
|||||
|
AAA1616614
|
Thermo Scientific Chemicals
A1616614 |
25 g |
Each for $233.29
|
|
|||||
|
AAA1616622
|
Thermo Scientific Chemicals
A1616622 |
100 g |
Each for $593.67
|
|
|||||
Description
L-(+)-Rhamnose monohydrate is used as a starting material for the production of furanones. It is an important material involved in the reaction of flavors developed during the preparation of various foods like bread, grilled meats, etc. It is also essential to test the permeability of the intestinal tract. Further, it is used as sweeting agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsL-(+)-Rhamnose monohydrate is used as a starting material for the production of furanones. It is an important material involved in the reaction of flavors developed during the preparation of various foods like bread, grilled meats, etc. It is also essential to test the permeability of the intestinal tract. Further, it is used as sweeting agent.
Solubility
Soluble in water and methanol. Slightly soluble in ethanol.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Chemical Identifiers
| 10030-85-0 | |
| 164.16 | |
| SHZGCJCMOBCMKK-HGVZOGFYSA-N | |
| 20849066 |
| C6H12O5 | |
| MFCD00149363,MFCD00136036 | |
| l-+-rhamnose monohydrate, 2r,3r,4s,5s-2,3,4,5-tetrahydroxyhexanal hydrate, l + rhamnopyranose, l-mannose, 6-deoxy-, monohydrate, 6-deoxy-l-mannose hydrate, rhamnose hydrate, l-rha hydrate, l-rhamnose hydrate, l +-rhamnose hydrate, a-l-rhamnose monohydrate | |
| C[C@@H]1O[C@@H](O)[C@H](O)[C@H](O)[C@H]1O |
Specifications
| 10030-85-0 | |
| 1.47 | |
| MFCD00149363,MFCD00136036 | |
| 5988592 | |
| l-+-rhamnose monohydrate, 2r,3r,4s,5s-2,3,4,5-tetrahydroxyhexanal hydrate, l + rhamnopyranose, l-mannose, 6-deoxy-, monohydrate, 6-deoxy-l-mannose hydrate, rhamnose hydrate, l-rha hydrate, l-rhamnose hydrate, l +-rhamnose hydrate, a-l-rhamnose monohydrate | |
| SHZGCJCMOBCMKK-HGVZOGFYSA-N | |
| C[C@@H]1O[C@@H](O)[C@H](O)[C@H](O)[C@H]1O | |
| 164.16 | |
| 182.19 (164.17 Anhydrous) | |
| L-(+)-Rhamnose monohydrate |
| 89°C to 94°C | |
| C6H12O5 | |
| 5 g | |
| 14,8172 | |
| Soluble in water and methanol. Slightly soluble in ethanol. | |
| +8° (c=10 in Water, 20h) | |
| (2R,3R,4S,5S)-2,3,4,5-tetrahydroxyhexanal;hydrate | |
| 20849066 | |
| 99% |
Safety and Handling
EINECSNumber : 222-793-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only