missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-(+)-Rhamnose monohydrate, 98+%

CAS: 10030-85-0 | C6H12O5 | 164.16 g/mol

Supplier:  Thermo Scientific Chemicals A1616614

 View more versions of this product

Catalog No. AAA1616614


Only null left

Description

Description

L-(+)-Rhamnose monohydrate is used as a starting material for the production of furanones. It is an important material involved in the reaction of flavors developed during the preparation of various foods like bread, grilled meats, etc. It is also essential to test the permeability of the intestinal tract. Further, it is used as sweeting agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-(+)-Rhamnose monohydrate is used as a starting material for the production of furanones. It is an important material involved in the reaction of flavors developed during the preparation of various foods like bread, grilled meats, etc. It is also essential to test the permeability of the intestinal tract. Further, it is used as sweeting agent.

Solubility
Soluble in water and methanol. Slightly soluble in ethanol.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Specifications

Specifications

L-(+)-Rhamnose monohydrate
89°C to 94°C
C6H12O5
25 g
14,8172
Soluble in water and methanol. Slightly soluble in ethanol.
+8° (c=10 in Water, 20h)
(2R,3R,4S,5S)-2,3,4,5-tetrahydroxyhexanal;hydrate
20849066
99%
10030-85-0
1.47
MFCD00149363,MFCD00136036
5988592
l-+-rhamnose monohydrate, 2r,3r,4s,5s-2,3,4,5-tetrahydroxyhexanal hydrate, l + rhamnopyranose, l-mannose, 6-deoxy-, monohydrate, 6-deoxy-l-mannose hydrate, rhamnose hydrate, l-rha hydrate, l-rhamnose hydrate, l +-rhamnose hydrate, a-l-rhamnose monohydrate
SHZGCJCMOBCMKK-HGVZOGFYSA-N
C[C@@H]1O[C@@H](O)[C@H](O)[C@H](O)[C@H]1O
164.16
182.19 (164.17 Anhydrous)

Chemical Identifiers

10030-85-0
164.16
SHZGCJCMOBCMKK-HGVZOGFYSA-N
20849066
C6H12O5
MFCD00149363,MFCD00136036
l-+-rhamnose monohydrate, 2r,3r,4s,5s-2,3,4,5-tetrahydroxyhexanal hydrate, l + rhamnopyranose, l-mannose, 6-deoxy-, monohydrate, 6-deoxy-l-mannose hydrate, rhamnose hydrate, l-rha hydrate, l-rhamnose hydrate, l +-rhamnose hydrate, a-l-rhamnose monohydrate
C[C@@H]1O[C@@H](O)[C@H](O)[C@H](O)[C@H]1O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 222-793-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only