missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Proline tert-Butyl Ester Hydrochloride 98.0+%, TCI America™
Supplier: TCI America P172925G
Specifications
| L-Proline tert-Butyl Ester Hydrochloride | |
| 112°C | |
| C9H18ClNO2 | |
| 25 g | |
| IUUYANMOEMBTBV-FJXQXJEOSA-N | |
| tert-butyl (2S)-pyrrolidine-2-carboxylate;hydrochloride | |
| 6453107 | |
| ≥98.0% (N,T) |
| 5497-76-7 | |
| White | |
| MFCD00153459 | |
| h-pro-otbu.hcl, l-proline tert-butyl ester hydrochloride, tert-butyl l-prolinate hydrochloride, l-proline tert-butyl ester hcl, pro-otbu hcl, h-pro-otbu hcl, h-pro-otbu hydrochloride, l-proline t-butyl ester hydrochloride, tert-butyl 2s-pyrrolidine-2-carboxylate hydrochloride, pro-otbu.hcl | |
| CC(C)(C)OC(=O)C1CCCN1.Cl | |
| 207.698 | |
| 207.70 | |
| Crystalline Powder |
Chemical Identifiers
| 5497-76-7 | |
| 207.698 | |
| IUUYANMOEMBTBV-FJXQXJEOSA-N | |
| 6453107 | |
| CC(C)(C)OC(=O)C1CCCN1.Cl |
| C9H18ClNO2 | |
| MFCD00153459 | |
| h-pro-otbu.hcl, l-proline tert-butyl ester hydrochloride, tert-butyl l-prolinate hydrochloride, l-proline tert-butyl ester hcl, pro-otbu hcl, h-pro-otbu hcl, h-pro-otbu hydrochloride, l-proline t-butyl ester hydrochloride, tert-butyl 2s-pyrrolidine-2-carboxylate hydrochloride, pro-otbu.hcl | |
| tert-butyl (2S)-pyrrolidine-2-carboxylate;hydrochloride |
Safety and Handling
TSCA : No