accessibility menu, dialog, popup

UserName

CAS RN 5497-76-7

ClCH3H3CCH3OO(S)HNH

CAS RN 5497-76-7

IUPAC Name: hydrogen tert-butyl (2S)-pyrrolidine-2-carboxylate chloride
Synonyms: hydrogen tert-butyl (2S)-pyrrolidine-2-carboxylate chloride
Molecular Weight (g/mol): 207.7
Molecular Formula: C9H18ClNO2
InChi Key: IUUYANMOEMBTBV-FJXQXJEOSA-N
SMILES: [H+].[Cl-].CC(C)(C)OC(=O)[C@@H]1CCCN1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)