missing translation for 'onlineSavingsMsg'
Learn More

L-Proline tert-Butyl Ester Hydrochloride 98.0+%, TCI America™

Catalog No. p-7135230
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P17295G 5 g
P172925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P17295G Supplier TCI America Supplier No. P17295G
Only null left

Chemical Identifiers

CAS 5497-76-7
Molecular Formula C9H18ClNO2
Molecular Weight (g/mol) 207.698
MDL Number MFCD00153459
InChI Key IUUYANMOEMBTBV-FJXQXJEOSA-N
Synonym h-pro-otbu.hcl, l-proline tert-butyl ester hydrochloride, tert-butyl l-prolinate hydrochloride, l-proline tert-butyl ester hcl, pro-otbu hcl, h-pro-otbu hcl, h-pro-otbu hydrochloride, l-proline t-butyl ester hydrochloride, tert-butyl 2s-pyrrolidine-2-carboxylate hydrochloride, pro-otbu.hcl
PubChem CID 6453107
IUPAC Name tert-butyl (2S)-pyrrolidine-2-carboxylate;hydrochloride
SMILES CC(C)(C)OC(=O)C1CCCN1.Cl

Specifications

Melting Point 112°C
Color White
Quantity 5 g
Formula Weight 207.70
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material L-Proline tert-Butyl Ester Hydrochloride