missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Phenylalanine, >99%, MP Biomedicals
Supplier: MP Biomedicals Inc 0219472325
Specifications
| L-Phenylalanine, >99% | |
| C9H11NO2 | |
| COLNVLDHVKWLRT-UHFFFAOYNA-N | |
| 2-amino-3-phenylpropanoic acid | |
| 165.19 | |
| CHEBI:17295 | |
| Cell Culture |
| 63-91-2 | |
| l-phenylalanine, phenylalanine, s-2-amino-3-phenylpropanoic acid, 3-phenyl-l-alanine, s-phenylalanine, 3-phenylalanine, 2s-2-amino-3-phenylpropanoic acid, s-2-amino-3-phenylpropionic acid, beta-phenyl-l-alanine, h-phe-oh | |
| NC(CC1=CC=CC=C1)C(O)=O | |
| 25 g | |
| 6140 | |
| 165.2 |
Chemical Identifiers
| 63-91-2 | |
| 165.19 | |
| l-phenylalanine, phenylalanine, s-2-amino-3-phenylpropanoic acid, 3-phenyl-l-alanine, s-phenylalanine, 3-phenylalanine, 2s-2-amino-3-phenylpropanoic acid, s-2-amino-3-phenylpropionic acid, beta-phenyl-l-alanine, h-phe-oh | |
| CHEBI:17295 | |
| NC(CC1=CC=CC=C1)C(O)=O |
| C9H11NO2 | |
| COLNVLDHVKWLRT-UHFFFAOYNA-N | |
| 6140 | |
| 2-amino-3-phenylpropanoic acid |
Safety and Handling
Recommended Storage : Room temperature