missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Phenylalanine, >99%, MP Biomedicals
$78.97 - $78.97
Chemical Identifiers
| CAS | 63-91-2 |
|---|---|
| Molecular Formula | C9H11NO2 |
| Molecular Weight (g/mol) | 165.19 |
| InChI Key | COLNVLDHVKWLRT-UHFFFAOYNA-N |
| Synonym | l-phenylalanine, phenylalanine, s-2-amino-3-phenylpropanoic acid, 3-phenyl-l-alanine, s-phenylalanine, 3-phenylalanine, 2s-2-amino-3-phenylpropanoic acid, s-2-amino-3-phenylpropionic acid, beta-phenyl-l-alanine, h-phe-oh |
| PubChem CID | 6140 |
| ChEBI | CHEBI:17295 |
| IUPAC Name | 2-amino-3-phenylpropanoic acid |
| SMILES | NC(CC1=CC=CC=C1)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN19472325
|
MP Biomedicals Inc
0219472325 |
25 g |
Each for $78.97
|
|
|||||
Chemical Identifiers
| 63-91-2 | |
| 165.19 | |
| l-phenylalanine, phenylalanine, s-2-amino-3-phenylpropanoic acid, 3-phenyl-l-alanine, s-phenylalanine, 3-phenylalanine, 2s-2-amino-3-phenylpropanoic acid, s-2-amino-3-phenylpropionic acid, beta-phenyl-l-alanine, h-phe-oh | |
| CHEBI:17295 | |
| NC(CC1=CC=CC=C1)C(O)=O |
| C9H11NO2 | |
| COLNVLDHVKWLRT-UHFFFAOYNA-N | |
| 6140 | |
| 2-amino-3-phenylpropanoic acid |
Specifications
| 63-91-2 | |
| l-phenylalanine, phenylalanine, s-2-amino-3-phenylpropanoic acid, 3-phenyl-l-alanine, s-phenylalanine, 3-phenylalanine, 2s-2-amino-3-phenylpropanoic acid, s-2-amino-3-phenylpropionic acid, beta-phenyl-l-alanine, h-phe-oh | |
| NC(CC1=CC=CC=C1)C(O)=O | |
| 25 g | |
| 6140 | |
| 165.2 | |
| L-Phenylalanine, >99% |
| C9H11NO2 | |
| COLNVLDHVKWLRT-UHFFFAOYNA-N | |
| 2-amino-3-phenylpropanoic acid | |
| 165.19 | |
| CHEBI:17295 | |
| Cell Culture |
Safety and Handling
Recommended Storage : Room temperature