missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Methionine Sulfoximine, 98-102%, Spectrum™ Chemical
M3114, 15985-39-4, C5H12N2O3S
$212.11 - $812.09
Chemical Identifiers
| CAS | 15985-39-4 |
|---|---|
| Molecular Formula | C5H12N2O3S |
| Molecular Weight (g/mol) | 180.22 |
| MDL Number | MFCD00002621 |
| InChI Key | SXTAYKAGBXMACB-DPVSGNNYSA-N |
| IUPAC Name | (2S)-2-azaniumyl-4-[imino(methyl)oxo-λâ¶-sulfanyl]butanoate |
| SMILES | CS(=N)(=O)CC[C@H]([NH3+])C([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18603950
|
Spectrum Chemical Mfg Cor
M3114100MG |
100 mg | Amber Glass Bottle |
Each for $212.11
|
|
||||
|
18603951
|
Spectrum Chemical Mfg Cor
M3114500MG |
500 mg | Amber Glass Bottle |
Each for $546.69
|
|
||||
|
18603952
|
Spectrum Chemical Mfg Cor
M31141GM |
1 g | Amber Glass Bottle |
Each for $812.09
|
|
||||
Description
Spectrum™ Chemical L-Methionine Sulfoximine , also known as 2-amino-4-(methylthio)butanoic acid, is an α-amino acid and nonpolar; it is classified as an essential amino acid in all metazoa. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.Chemical Identifiers
| 15985-39-4 | |
| 180.22 | |
| SXTAYKAGBXMACB-DPVSGNNYSA-N | |
| CS(=N)(=O)CC[C@H]([NH3+])C([O-])=O |
| C5H12N2O3S | |
| MFCD00002621 | |
| (2S)-2-azaniumyl-4-[imino(methyl)oxo-λâ¶-sulfanyl]butanoate |
Specifications
| 15985-39-4 | |
| Amber Glass Bottle | |
| MFCD00002621 | |
| +12° t0 +15° | |
| CS(=N)(=O)CC[C@H]([NH3+])C([O-])=O | |
| 180.22 | |
| Reagent |
| 100% | |
| C5H12N2O3S | |
| 100 mg | |
| SXTAYKAGBXMACB-DPVSGNNYSA-N | |
| (2S)-2-azaniumyl-4-[imino(methyl)oxo-λâ¶-sulfanyl]butanoate | |
| 98 to 102% |