missing translation for 'onlineSavingsMsg'
Learn More

L-Methionine Sulfoximine, 98-102%, Spectrum™ Chemical

Catalog No. 18603952
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603952 1 g Amber Glass Bottle
18603950 100 mg Amber Glass Bottle
18603951 500 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18603952 Supplier Spectrum Chemical Mfg Cor Supplier No. M31141GM
Get Up to $500 in Chemicals with Purchase
View all promotions

M3114, 15985-39-4, C5H12N2O3S

Spectrum™ Chemical L-Methionine Sulfoximine , also known as 2-amino-4-(methylthio)butanoic acid, is an α-amino acid and nonpolar; it is classified as an essential amino acid in all metazoa. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 15985-39-4
Molecular Formula C5H12N2O3S
Molecular Weight (g/mol) 180.22
MDL Number MFCD00002621
InChI Key SXTAYKAGBXMACB-DPVSGNNYSA-N
IUPAC Name (2S)-2-azaniumyl-4-[imino(methyl)oxo-λ⁶-sulfanyl]butanoate
SMILES CS(=N)(=O)CC[C@H]([NH3+])C([O-])=O

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 1 g
Specific Rotation +12° t0 +15°
Percent Purity 98 to 102%
Grade Reagent