missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Methionine Sulfoximine, 98-102%, Spectrum™ Chemical
M3114, 15985-39-4, C5H12N2O3S
Supplier: Spectrum Chemical Mfg Cor M3114500MG
Description
Spectrum™ Chemical L-Methionine Sulfoximine , also known as 2-amino-4-(methylthio)butanoic acid, is an α-amino acid and nonpolar; it is classified as an essential amino acid in all metazoa. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.Specifications
| 15985-39-4 | |
| Amber Glass Bottle | |
| MFCD00002621 | |
| +12° t0 +15° | |
| CS(=N)(=O)CC[C@H]([NH3+])C([O-])=O | |
| 180.22 | |
| Reagent |
| 100% | |
| C5H12N2O3S | |
| 500 mg | |
| SXTAYKAGBXMACB-DPVSGNNYSA-N | |
| (2S)-2-azaniumyl-4-[imino(methyl)oxo-λâ¶-sulfanyl]butanoate | |
| 98 to 102% |
Chemical Identifiers
| 15985-39-4 | |
| 180.22 | |
| SXTAYKAGBXMACB-DPVSGNNYSA-N | |
| CS(=N)(=O)CC[C@H]([NH3+])C([O-])=O |
| C5H12N2O3S | |
| MFCD00002621 | |
| (2S)-2-azaniumyl-4-[imino(methyl)oxo-λâ¶-sulfanyl]butanoate |