missing translation for 'onlineSavingsMsg'
Learn More

L-Methionine methyl ester hydrochloride, 99%

CAS: 2491-18-1 | C6H14ClNO2S | 199.69 g/mol

$70.08 - $631.97

Chemical Identifiers

CAS 2491-18-1
Molecular Formula C6H14ClNO2S
Molecular Weight (g/mol) 199.69
MDL Number MFCD00012491,MFCD00067540
InChI Key MEVUPUNLVKELNV-UHFFFAOYNA-N
Synonym h-met-ome.hcl, l-methionine methyl ester hydrochloride, h-met-ome hydrochloride, methionine methyl ester hcl, methyl 2s-2-amino-4-methylsulfanyl butanoate hydrochloride, h-d-met-ome hydrochloride, met-ome.hcl, met-ome. hcl, h-met-ome. hcl, pubchem14925
PubChem CID 11435579
IUPAC Name methyl (2S)-2-amino-4-methylsulfanylbutanoate;hydrochloride
SMILES [H+].[Cl-].COC(=O)C(N)CCSC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1595206
View Documents
Thermo Scientific Chemicals
A1595206
5 g
Each for $70.08
Only null left
 
AAA1595214
View Documents
Thermo Scientific Chemicals
A1595214
25 g
Each for $217.14
Only null left
 
AAA1595222
View Documents
Thermo Scientific Chemicals
A1595222
100 g
Each for $631.97
Only null left
 
Description

Description

L-Methionine methyl ester hydrochloride is used as a starting material for the synthesis of a protected glycine derivative, a versatile asymmetric building block.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Methionine methyl ester hydrochloride is used as a starting material for the synthesis of a protected glycine derivative, a versatile asymmetric building block.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

2491-18-1
199.69
MEVUPUNLVKELNV-UHFFFAOYNA-N
11435579
[H+].[Cl-].COC(=O)C(N)CCSC
C6H14ClNO2S
MFCD00012491,MFCD00067540
h-met-ome.hcl, l-methionine methyl ester hydrochloride, h-met-ome hydrochloride, methionine methyl ester hcl, methyl 2s-2-amino-4-methylsulfanyl butanoate hydrochloride, h-d-met-ome hydrochloride, met-ome.hcl, met-ome. hcl, h-met-ome. hcl, pubchem14925
methyl (2S)-2-amino-4-methylsulfanylbutanoate;hydrochloride

Specifications

151°C to 152°C
C6H14ClNO2S
3913214
Soluble in water.
[H+].[Cl-].COC(=O)C(N)CCSC
5 g
11435579
199.7
99%
2491-18-1
MFCD00012491,MFCD00067540
h-met-ome.hcl, l-methionine methyl ester hydrochloride, h-met-ome hydrochloride, methionine methyl ester hcl, methyl 2s-2-amino-4-methylsulfanyl butanoate hydrochloride, h-d-met-ome hydrochloride, met-ome.hcl, met-ome. hcl, h-met-ome. hcl, pubchem14925
MEVUPUNLVKELNV-UHFFFAOYNA-N
methyl (2S)-2-amino-4-methylsulfanylbutanoate;hydrochloride
199.69
+26° (c=5 in Water)
Hygroscopic
L-Methionine methyl ester hydrochloride
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 219-651-9

TSCA : No

Recommended Storage : Store at -20°C

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only