missing translation for 'onlineSavingsMsg'
Learn More

L-Methionine methyl ester hydrochloride, 99%

CAS: 2491-18-1 | C6H14ClNO2S | 199.69 g/mol

Supplier:  Thermo Scientific Chemicals A1595222

 View more versions of this product

Catalog No. AAA1595222


Only null left
Explore available promotions

Description

Description

L-Methionine methyl ester hydrochloride is used as a starting material for the synthesis of a protected glycine derivative, a versatile asymmetric building block.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Methionine methyl ester hydrochloride is used as a starting material for the synthesis of a protected glycine derivative, a versatile asymmetric building block.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

151°C to 152°C
2491-18-1
MFCD00012491,MFCD00067540
h-met-ome.hcl, l-methionine methyl ester hydrochloride, h-met-ome hydrochloride, methionine methyl ester hcl, methyl 2s-2-amino-4-methylsulfanyl butanoate hydrochloride, h-d-met-ome hydrochloride, met-ome.hcl, met-ome. hcl, h-met-ome. hcl, pubchem14925
MEVUPUNLVKELNV-UHFFFAOYNA-N
methyl (2S)-2-amino-4-methylsulfanylbutanoate;hydrochloride
199.69
+26° (c=5 in Water)
Hygroscopic
L-Methionine methyl ester hydrochloride
C6H14ClNO2S
3913214
Soluble in water.
[H+].[Cl-].COC(=O)C(N)CCSC
100 g
11435579
199.7
99%

Chemical Identifiers

2491-18-1
199.69
MEVUPUNLVKELNV-UHFFFAOYNA-N
11435579
[H+].[Cl-].COC(=O)C(N)CCSC
C6H14ClNO2S
MFCD00012491,MFCD00067540
h-met-ome.hcl, l-methionine methyl ester hydrochloride, h-met-ome hydrochloride, methionine methyl ester hcl, methyl 2s-2-amino-4-methylsulfanyl butanoate hydrochloride, h-d-met-ome hydrochloride, met-ome.hcl, met-ome. hcl, h-met-ome. hcl, pubchem14925
methyl (2S)-2-amino-4-methylsulfanylbutanoate;hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 219-651-9

TSCA : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only