accessibility menu, dialog, popup

UserName

H3CSNH2(S)OOCH3HCl

CAS RN 2491-18-1

IUPAC Name: hydrogen methyl 2-amino-4-(methylsulfanyl)butanoate chloride
Synonyms: hydrogen methyl 2-amino-4-(methylsulfanyl)butanoate chloride
Molecular Weight (g/mol): 199.69
Molecular Formula: C6H14ClNO2S
InChi Key: MEVUPUNLVKELNV-UHFFFAOYNA-N
SMILES: [H+].[Cl-].COC(=O)C(N)CCSC
Percent Purity 
  • (1)
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (5)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)