missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-(+)-Lyxose 98.0+%, TCI America™
$311.38 - $1,070.82
Chemical Identifiers
| CAS | 1949-78-6 |
|---|---|
| Molecular Formula | C5H10O5 |
| Molecular Weight (g/mol) | 150.13 |
| MDL Number | MFCD00064111 |
| InChI Key | PYMYPHUHKUWMLA-YUPRTTJUSA-N |
| Synonym | l-lyxose, aldehydo-l-lyxose, lyxose, l, l-lyx, 2r,3r,4s-2,3,4,5-tetrahydroxypentanal, unii-m7j5hm9dy4, m7j5hm9dy4, l +-lyxose, l-lyxose 9ci |
| PubChem CID | 644176 |
| ChEBI | CHEBI:28480 |
| IUPAC Name | (2R,3R,4S)-2,3,4,5-tetrahydroxypentanal |
| SMILES | C(C(C(C(C=O)O)O)O)O |
Chemical Identifiers
| C5H10O5 | |
| MFCD00064111 | |
| l-lyxose, aldehydo-l-lyxose, lyxose, l, l-lyx, 2r,3r,4s-2,3,4,5-tetrahydroxypentanal, unii-m7j5hm9dy4, m7j5hm9dy4, l +-lyxose, l-lyxose 9ci | |
| CHEBI:28480 | |
| C(C(C(C(C=O)O)O)O)O |
Specifications
| 1949-78-6 | |
| C5H10O5 | |
| l-lyxose, aldehydo-l-lyxose, lyxose, l, l-lyx, 2r,3r,4s-2,3,4,5-tetrahydroxypentanal, unii-m7j5hm9dy4, m7j5hm9dy4, l +-lyxose, l-lyxose 9ci | |
| C(C(C(C(C=O)O)O)O)O | |
| 150.13 | |
| CHEBI:28480 | |
| ≥98.0% (HPLC) | |
| L-(+)-Lyxose |
| White | |
| MFCD00064111 | |
| PYMYPHUHKUWMLA-YUPRTTJUSA-N | |
| (2R,3R,4S)-2,3,4,5-tetrahydroxypentanal | |
| 644176 | |
| 150.13 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes