missing translation for 'onlineSavingsMsg'
Learn More

L-(+)-Lyxose 98.0+%, TCI America™

Catalog No. L01535G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L01535G 5 g
L01531G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L01535G Supplier TCI America Supplier No. L01535G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1949-78-6
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13
MDL Number MFCD00064111
InChI Key PYMYPHUHKUWMLA-YUPRTTJUSA-N
Synonym l-lyxose, aldehydo-l-lyxose, lyxose, l, l-lyx, 2r,3r,4s-2,3,4,5-tetrahydroxypentanal, unii-m7j5hm9dy4, m7j5hm9dy4, l +-lyxose, l-lyxose 9ci
PubChem CID 644176
ChEBI CHEBI:28480
IUPAC Name (2R,3R,4S)-2,3,4,5-tetrahydroxypentanal
SMILES C(C(C(C(C=O)O)O)O)O

Specifications

Color White
Quantity 5 g
Formula Weight 150.13
Percent Purity ≥98.0% (HPLC)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material L-(+)-Lyxose