missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-(+)-Lysine Monohydrochloride 98.0+%, TCI America™
Chemical Identifiers
| CAS | 657-27-2 |
|---|---|
| Molecular Formula | C6H15ClN2O2 |
| Molecular Weight (g/mol) | 182.65 |
| MDL Number | MFCD00064564 MFCD00081870 |
| InChI Key | BVHLGVCQOALMSV-JEDNCBNOSA-N |
| Synonym | l-lysine hydrochloride, l-lysine monohydrochloride, lysine hydrochloride, h-lys-oh.hcl, l-lysine, monohydrochloride, lyamine, darvyl, lysion, s-2,6-diaminohexanoic acid hydrochloride, enisyl |
| PubChem CID | 69568 |
| ChEBI | CHEBI:53633 |
| IUPAC Name | (2S)-2,6-diaminohexanoic acid hydrochloride |
| SMILES | Cl.NCCCC[C@H](N)C(O)=O |
Chemical Identifiers
| C6H15ClN2O2 | |
| MFCD00064564 MFCD00081870 | |
| l-lysine hydrochloride, l-lysine monohydrochloride, lysine hydrochloride, h-lys-oh.hcl, l-lysine, monohydrochloride, lyamine, darvyl, lysion, s-2,6-diaminohexanoic acid hydrochloride, enisyl | |
| CHEBI:53633 | |
| Cl.NCCCC[C@H](N)C(O)=O |
Specifications
| 657-27-2 | |
| C6H15ClN2O2 | |
| l-lysine hydrochloride, l-lysine monohydrochloride, lysine hydrochloride, h-lys-oh.hcl, l-lysine, monohydrochloride, lyamine, darvyl, lysion, s-2,6-diaminohexanoic acid hydrochloride, enisyl | |
| Cl.NCCCC[C@H](N)C(O)=O | |
| 182.65 | |
| CHEBI:53633 | |
| ≥98.0% (T) | |
| L-(+)-Lysine Monohydrochloride |
| White | |
| MFCD00064564 MFCD00081870 | |
| BVHLGVCQOALMSV-JEDNCBNOSA-N | |
| (2S)-2,6-diaminohexanoic acid hydrochloride | |
| 69568 | |
| 182.65 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : OL5650000
TSCA : Yes