missing translation for 'onlineSavingsMsg'
Learn More

L-(+)-Lysine Monohydrochloride 98.0+%, TCI America™

Catalog No. L007125G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L007125G 25 g
L0071500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L007125G Supplier TCI America Supplier No. L007125G

Chemical Identifiers

CAS 657-27-2
Molecular Formula C6H15ClN2O2
Molecular Weight (g/mol) 182.65
MDL Number MFCD00064564 MFCD00081870
InChI Key BVHLGVCQOALMSV-JEDNCBNOSA-N
Synonym l-lysine hydrochloride, l-lysine monohydrochloride, lysine hydrochloride, h-lys-oh.hcl, l-lysine, monohydrochloride, lyamine, darvyl, lysion, s-2,6-diaminohexanoic acid hydrochloride, enisyl
PubChem CID 69568
ChEBI CHEBI:53633
IUPAC Name (2S)-2,6-diaminohexanoic acid hydrochloride
SMILES Cl.NCCCC[C@H](N)C(O)=O

Specifications

Color White
Quantity 25 g
Formula Weight 182.65
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-(+)-Lysine Monohydrochloride
RTECSNumber OL5650000
TSCA Yes