missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals L-Lysine monohydrochloride, Cell Culture Reagent
CAS: 657-27-2 | C6H15ClN2O2 | 182.65 g/mol
Chemical Identifiers
| CAS | 657-27-2 |
|---|---|
| Molecular Formula | C6H15ClN2O2 |
| Molecular Weight (g/mol) | 182.65 |
| MDL Number | MFCD00064564 MFCD00081870 |
| InChI Key | BVHLGVCQOALMSV-JEDNCBNOSA-N |
| Synonym | l-lysine hydrochloride, l-lysine monohydrochloride, lysine hydrochloride, h-lys-oh.hcl, l-lysine, monohydrochloride, lyamine, darvyl, lysion, s-2,6-diaminohexanoic acid hydrochloride, enisyl |
| PubChem CID | 69568 |
| ChEBI | CHEBI:53633 |
| IUPAC Name | (2S)-2,6-diaminohexanoic acid;hydrochloride |
| SMILES | Cl.NCCCC[C@H](N)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAJ6209922
|
Thermo Scientific Chemicals
J6209922 |
100 g |
N/A
|
|
|||||
|
AAJ6209936
|
Thermo Scientific Chemicals
J6209936 |
500 g |
N/A
|
|
|||||
|
AAJ62099A1
|
Thermo Scientific Chemicals
J62099A1 |
1 kg |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| C6H15ClN2O2 | |
| MFCD00064564 MFCD00081870 | |
| l-lysine hydrochloride, l-lysine monohydrochloride, lysine hydrochloride, h-lys-oh.hcl, l-lysine, monohydrochloride, lyamine, darvyl, lysion, s-2,6-diaminohexanoic acid hydrochloride, enisyl | |
| CHEBI:53633 | |
| Cl.NCCCC[C@H](N)C(O)=O |
Specifications
| 657-27-2 | |
| C6H15ClN2O2 | |
| 3563889 | |
| l-lysine hydrochloride, l-lysine monohydrochloride, lysine hydrochloride, h-lys-oh.hcl, l-lysine, monohydrochloride, lyamine, darvyl, lysion, s-2,6-diaminohexanoic acid hydrochloride, enisyl | |
| Cl.NCCCC[C@H](N)C(O)=O | |
| 182.65 | |
| CHEBI:53633 | |
| Cell Culture Reagent | |
| L-Lysine monohydrochloride |
| White | |
| MFCD00064564 MFCD00081870 | |
| 14,5636 | |
| BVHLGVCQOALMSV-JEDNCBNOSA-N | |
| (2S)-2,6-diaminohexanoic acid;hydrochloride | |
| 69568 | |
| 182.65 | |
| Powder |
Safety and Handling
EINECSNumber : 211-519-9
RTECSNumber : OL5650000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only