missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Lysine monohydrochloride, Cell Culture Reagent

Catalog No. AAJ6209922
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6209922 100 g
AAJ6209936 500 g
AAJ62099A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6209922 Supplier Thermo Scientific Chemicals Supplier No. J6209922

CAS: 657-27-2 | C6H15ClN2O2 | 182.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 657-27-2
Molecular Formula C6H15ClN2O2
Molecular Weight (g/mol) 182.65
MDL Number MFCD00064564 MFCD00081870
InChI Key BVHLGVCQOALMSV-JEDNCBNOSA-N
Synonym l-lysine hydrochloride, l-lysine monohydrochloride, lysine hydrochloride, h-lys-oh.hcl, l-lysine, monohydrochloride, lyamine, darvyl, lysion, s-2,6-diaminohexanoic acid hydrochloride, enisyl
PubChem CID 69568
ChEBI CHEBI:53633
IUPAC Name (2S)-2,6-diaminohexanoic acid;hydrochloride
SMILES Cl.NCCCC[C@H](N)C(O)=O

Specifications

Color White
Beilstein 3563889
Merck Index 14,5636
Quantity 100 g
Formula Weight 182.65
Grade Cell Culture Reagent
Physical Form Powder
Chemical Name or Material L-Lysine monohydrochloride
EINECSNumber 211-519-9
RTECSNumber OL5650000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only