missing translation for 'onlineSavingsMsg'
Learn More

L-Lysine Monohydrochloride, 98.5-101.5%, Spectrum™ Chemical

Chemical Identifiers

CAS 657-27-2
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
18611365
View Documents Promotion Details
N/A 100 g Poly Pail N/A
Only null left
 
18611367
View Documents Promotion Details
N/A 500 g Poly Pail N/A
Only null left
 
18611363
View Documents Promotion Details
N/A 1 kg Poly Pail N/A
Only null left
 
18611364
View Documents Promotion Details
N/A 5 kg Poly Pail N/A
Only null left
 
18611366
View Documents Promotion Details
N/A 25 kg Poly Pail N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

657-27-2

Specifications

657-27-2
1
3e-05
0.002
98.5 to 101.5%
L-Lysine Monohydrochloride
1.5e-06
1e-05
0.01
+20.3° to +21.3°
Reagent
Videos
SDS
Documents

Documents

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.