missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Lysine Monohydrochloride, 98.5-101.5%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor LY1055KG
| Quantity | 5 kg |
|---|---|
| Packaging | Poly Pail |
Chemical Identifiers
| 657-27-2 |
Specifications
| L-Lysine Monohydrochloride | |
| 1.5e-06 | |
| 1e-05 | |
| 0.01 | |
| 0.002 | |
| 5 kg | |
| Reagent |
| 657-27-2 | |
| 1 | |
| 3e-05 | |
| Poly Pail | |
| +20.3° to +21.3° | |
| 98.5 to 101.5% |