missing translation for 'onlineSavingsMsg'
Learn More

L-Lysine, Monochloride, Monohydrate, USP, EP, bioCERTIFIED, 98.5-101.5%, Spectrum™ Chemical

Spectrum™ Chemical bioCERTIFIED products are suitable for biopharmaceutical production.

Chemical Identifiers

CAS 657-27-2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
18614858
View Documents Promotion Details
N/A 1 kg Plastic Bottle N/A
Only null left
 
18614859
View Documents Promotion Details
N/A 12 kg Plastic Bottle N/A
Only null left
 
Description

Description

  • Full testing including endotoxin and elemental analysis
  • Comprehensive regulatory and scientific documentation
  • Change control and batch traceabilitiy
  • Biopharm industry standard CFR 210/211 cGMP
Specifications

Chemical Identifiers

657-27-2

Specifications

0.02%
6.0 EU/g
To pass test
1%
+20.8° to +21.5°
98.5 to 101.5%
L-Lysine Monohydrochloride
657-27-2
10 ppm
5 mg/kg
0.1%
Clear and Colorless: Not less than 98.0%
USP/JP/bioCERTIFIED
Videos
SDS
Documents

Documents