missing translation for 'onlineSavingsMsg'
Learn More

L-Lysine, Monochloride, Monohydrate, USP, EP, bioCERTIFIED, 98.5-101.5%, Spectrum™ Chemical

Catalog No. 18614858
Change view
Click to view available options
Quantity:
1 kg
12 kg
Packaging:
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614858 1 kg Plastic Bottle
18614859 12 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18614858 Supplier Spectrum Chemical Mfg Cor Supplier No. L98001KG

Spectrum™ Chemical bioCERTIFIED products are suitable for biopharmaceutical production.

  • Full testing including endotoxin and elemental analysis
  • Comprehensive regulatory and scientific documentation
  • Change control and batch traceabilitiy
  • Biopharm industry standard CFR 210/211 cGMP

Chemical Identifiers

CAS 657-27-2

Specifications

Ammonium 0.02%
Bacterial Endotoxins 6.0 EU/g
Heavy Metals (as Pb) 10 ppm
Identification To pass test
Lead (Pb) 5 mg/kg
Loss on Drying 1%
Packaging Plastic Bottle
Residue after Ignition 0.1%
Specific Rotation +20.8° to +21.5°
State of Solution (%T) Clear and Colorless: Not less than 98.0%
Quantity 1 kg
Percent Purity 98.5 to 101.5%
Grade USP/JP/bioCERTIFIED
Chemical Name or Material L-Lysine Monohydrochloride
Show More Show Less