missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Lysine methyl ester dihydrochloride, 98%, Thermo Scientific™
$56.31 - $400.71
Chemical Identifiers
| CAS | 26348-70-9 |
|---|---|
| Molecular Formula | C7H18Cl2N2O2 |
| Molecular Weight (g/mol) | 233.13 |
| MDL Number | MFCD00039067 |
| InChI Key | SXZCBVCQHOJXDR-ILKKLZGPSA-N |
| Synonym | l-lysine methyl ester dihydrochloride, h-lys-ome.2hcl, methyl l-lysinate dihydrochloride, l-lysine methyl ester 2hcl, s-methyl 2,6-diaminohexanoate dihydrochloride, l-lysine, methyl ester, dihydrochloride, methyl l-lysinate hcl, lysine, methyl ester, dihydrochloride, methyl 2s-2,6-diaminohexanoate dihydrochloride, h-lys-ome |
| PubChem CID | 117778 |
| IUPAC Name | methyl (2S)-2,6-diaminohexanoate;dihydrochloride |
| SMILES | [H+].[H+].[Cl-].[Cl-].COC(=O)[C@@H](N)CCCCN |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC459270050
|
N/A | 5g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC459270250
|
N/A | 25g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC459271000
|
N/A | 100g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 26348-70-9 | |
| 233.13 | |
| SXZCBVCQHOJXDR-ILKKLZGPSA-N | |
| 117778 | |
| [H+].[H+].[Cl-].[Cl-].COC(=O)[C@@H](N)CCCCN |
| C7H18Cl2N2O2 | |
| MFCD00039067 | |
| l-lysine methyl ester dihydrochloride, h-lys-ome.2hcl, methyl l-lysinate dihydrochloride, l-lysine methyl ester 2hcl, s-methyl 2,6-diaminohexanoate dihydrochloride, l-lysine, methyl ester, dihydrochloride, methyl l-lysinate hcl, lysine, methyl ester, dihydrochloride, methyl 2s-2,6-diaminohexanoate dihydrochloride, h-lys-ome | |
| methyl (2S)-2,6-diaminohexanoate;dihydrochloride |
Specifications
| 26348-70-9 | |
| C7H18Cl2N2O2 | |
| l-lysine methyl ester dihydrochloride, h-lys-ome.2hcl, methyl l-lysinate dihydrochloride, l-lysine methyl ester 2hcl, s-methyl 2,6-diaminohexanoate dihydrochloride, l-lysine, methyl ester, dihydrochloride, methyl l-lysinate hcl, lysine, methyl ester, dihydrochloride, methyl 2s-2,6-diaminohexanoate dihydrochloride, h-lys-ome | |
| SXZCBVCQHOJXDR-ILKKLZGPSA-N | |
| methyl (2S)-2,6-diaminohexanoate;dihydrochloride | |
| 117778 | |
| 98% |
| Authentic | |
| MFCD00039067 | |
| Solubility in water: soluble. | |
| [H+].[H+].[Cl-].[Cl-].COC(=O)[C@@H](N)CCCCN | |
| 233.13 | |
| 233.14 | |
| L-Lysine methyl ester dihydrochloride |
RUO – Research Use Only