missing translation for 'onlineSavingsMsg'
Learn More

L-Lysine methyl ester dihydrochloride, 98%

Catalog No. AC459270050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC459270050 5 g
AC459270250 25 g
AC459271000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC459270050 Supplier Thermo Scientific Chemicals Supplier No. 459270050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 26348-70-9 | C7H18Cl2N2O2 | 233.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 26348-70-9
Molecular Formula C7H18Cl2N2O2
Molecular Weight (g/mol) 233.13
MDL Number MFCD00039067
InChI Key SXZCBVCQHOJXDR-ILKKLZGPSA-N
Synonym l-lysine methyl ester dihydrochloride, h-lys-ome.2hcl, methyl l-lysinate dihydrochloride, l-lysine methyl ester 2hcl, s-methyl 2,6-diaminohexanoate dihydrochloride, l-lysine, methyl ester, dihydrochloride, methyl l-lysinate hcl, lysine, methyl ester, dihydrochloride, methyl 2s-2,6-diaminohexanoate dihydrochloride, h-lys-ome
PubChem CID 117778
IUPAC Name methyl (2S)-2,6-diaminohexanoate;dihydrochloride
SMILES [H+].[H+].[Cl-].[Cl-].COC(=O)[C@@H](N)CCCCN

Specifications

Melting Point 200.0°C
Infrared Spectrum Authentic
Solubility Information Solubility in water: soluble.
Quantity 5 g
Formula Weight 233.14
Percent Purity 98%
Chemical Name or Material L-Lysine methyl ester dihydrochloride

RUO – Research Use Only