missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Lysine methyl ester dihydrochloride, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 459270250
| Quantity | 25g |
|---|
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 26348-70-9 | |
| 233.13 | |
| SXZCBVCQHOJXDR-ILKKLZGPSA-N | |
| 117778 | |
| [H+].[H+].[Cl-].[Cl-].COC(=O)[C@@H](N)CCCCN |
| C7H18Cl2N2O2 | |
| MFCD00039067 | |
| l-lysine methyl ester dihydrochloride, h-lys-ome.2hcl, methyl l-lysinate dihydrochloride, l-lysine methyl ester 2hcl, s-methyl 2,6-diaminohexanoate dihydrochloride, l-lysine, methyl ester, dihydrochloride, methyl l-lysinate hcl, lysine, methyl ester, dihydrochloride, methyl 2s-2,6-diaminohexanoate dihydrochloride, h-lys-ome | |
| methyl (2S)-2,6-diaminohexanoate;dihydrochloride |
Specifications
| L-Lysine methyl ester dihydrochloride | |
| Authentic | |
| MFCD00039067 | |
| Solubility in water: soluble. | |
| [H+].[H+].[Cl-].[Cl-].COC(=O)[C@@H](N)CCCCN | |
| 233.13 | |
| 117778 | |
| 98% |
| 26348-70-9 | |
| C7H18Cl2N2O2 | |
| l-lysine methyl ester dihydrochloride, h-lys-ome.2hcl, methyl l-lysinate dihydrochloride, l-lysine methyl ester 2hcl, s-methyl 2,6-diaminohexanoate dihydrochloride, l-lysine, methyl ester, dihydrochloride, methyl l-lysinate hcl, lysine, methyl ester, dihydrochloride, methyl 2s-2,6-diaminohexanoate dihydrochloride, h-lys-ome | |
| SXZCBVCQHOJXDR-ILKKLZGPSA-N | |
| methyl (2S)-2,6-diaminohexanoate;dihydrochloride | |
| 25g | |
| 233.14 |