missing translation for 'onlineSavingsMsg'
Learn More

L-(+)-Lysine, Hydrate, 98.5-101.5%, Spectrum™ Chemical

$225.86 - $15574.32

Chemical Identifiers

CAS 39665-12-8
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18611171
View Documents
Spectrum Chemical Mfg Cor
L11465GM
5 g Amber Glass Bottle
Each for $225.86
Only null left
 
18611170
View Documents
Spectrum Chemical Mfg Cor
L114625GM
25 g Amber Glass Bottle
Each for $352.39
Only null left
 
18611169
View Documents
Spectrum Chemical Mfg Cor
L1146100GM
100 g Amber Glass Bottle
Each for $1,001.61
Only null left
 
18611168
View Documents
Spectrum Chemical Mfg Cor
L11461KGBL
1 kg Amber Glass Bottle
Each for $5,160.82
Only null left
 
18611172
View Documents
Spectrum Chemical Mfg Cor
L11465KGBL
5 kg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

39665-12-8

Specifications

200 ppm
1
300 ppm
10 ppm
Amber Glass Bottle
200 ppm
5 g
Reagent
39665-12-8
9.0 to 10.5
10 ppm
0.005
+25.5° to+27.0°
10.0 - 11.5%
98.5 to 101.5%
L-(+)-Lysine Hydrate
Videos
SDS
Documents

Documents

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.