missing translation for 'onlineSavingsMsg'
Learn More

L-(+)-Lysine, Hydrate, 98.5-101.5%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor L1146100GM

 View more versions of this product

Catalog No. 18611169


Only null left

Description

Description

Specifications

Specifications

L-(+)-Lysine Hydrate
39665-12-8
9.0 to 10.5
10 ppm
0.005
+25.5° to+27.0°
10.0 - 11.5%
98.5 to 101.5%
200 ppm
1
300 ppm
10 ppm
Amber Glass Bottle
200 ppm
100 g
Reagent

Chemical Identifiers

39665-12-8
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.