missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Lysine, 98%

CAS: 56-87-1 | C6H14N2O2 | 146.19 g/mol

Supplier:  Thermo Scientific Chemicals 303345000

 View more versions of this product

Catalog No. AC303345000


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

214.0°C to 216.0°C
56-87-1
Authentic
C6H14N2O2
MFCD00064433
l-lysine, lysine, h-lys-oh, lysine acid, s-lysine, aminutrin, 2s-2,6-diaminohexanoic acid, l-+-lysine, alpha-lysine, lysinum
KDXKERNSBIXSRK-MLHKIVSYNA-N
(2S)-2,6-diaminohexanoic acid
146.19
CHEBI:18019
98%
L-Lysine
White to Yellow
98%
Glass Bottle
+23.0° to +27.0° (20°C, 589nm) (C=2, 1N HCl)
3% max.
NCCCC[C@H](N)C(O)=O
500 g
5962
146.19
Powder or Crystals

Chemical Identifiers

56-87-1
146.19
KDXKERNSBIXSRK-MLHKIVSYNA-N
5962
(2S)-2,6-diaminohexanoic acid
C6H14N2O2
MFCD00064433
l-lysine, lysine, h-lys-oh, lysine acid, s-lysine, aminutrin, 2s-2,6-diaminohexanoic acid, l-+-lysine, alpha-lysine, lysinum
CHEBI:18019
NCCCC[C@H](N)C(O)=O
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only