accessibility menu, dialog, popup

UserName

H2N(S)NH2OHO

CAS RN 56-87-1

IUPAC Name: (2S)-2,6-diaminohexanoic acid
Synonyms: L-lysine
Molecular Weight (g/mol): 146.19
Molecular Formula: C6H14N2O2
InChi Key: KDXKERNSBIXSRK-MLHKIVSYNA-N
SMILES: NCCCC[C@H](N)C(O)=O
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (2)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (1)
  • (2)