missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Leucic Acid 99.0+%, TCI America™
$55.84 - $431.25
Chemical Identifiers
| CAS | 13748-90-8 |
|---|---|
| Molecular Formula | C6H12O3 |
| Molecular Weight (g/mol) | 132.159 |
| MDL Number | MFCD00064214 |
| InChI Key | LVRFTAZAXQPQHI-YFKPBYRVSA-N |
| Synonym | l-leucic acid, s-2-hydroxy-4-methylpentanoic acid, 2s-2-hydroxy-4-methylpentanoic acid, l-2-hydroxy-4-methylvaleric acid, s-leucic acid, s-2-hydroxy-4-methylvaleric acid, l-2-hydroxyisocaproic acid, l-alpha-hydroxyisocaproic acid, s---2-hydroxyisocaproic acid, pentanoic acid, 2-hydroxy-4-methyl-, 2s |
| PubChem CID | 83697 |
| ChEBI | CHEBI:44510 |
| IUPAC Name | (2S)-2-hydroxy-4-methylpentanoic acid |
| SMILES | CC(C)CC(C(=O)O)O |
Chemical Identifiers
| 13748-90-8 | |
| 132.159 | |
| LVRFTAZAXQPQHI-YFKPBYRVSA-N | |
| 83697 | |
| (2S)-2-hydroxy-4-methylpentanoic acid |
| C6H12O3 | |
| MFCD00064214 | |
| l-leucic acid, s-2-hydroxy-4-methylpentanoic acid, 2s-2-hydroxy-4-methylpentanoic acid, l-2-hydroxy-4-methylvaleric acid, s-leucic acid, s-2-hydroxy-4-methylvaleric acid, l-2-hydroxyisocaproic acid, l-alpha-hydroxyisocaproic acid, s---2-hydroxyisocaproic acid, pentanoic acid, 2-hydroxy-4-methyl-, 2s | |
| CHEBI:44510 | |
| CC(C)CC(C(=O)O)O |
Specifications
| 13748-90-8 | |
| White | |
| MFCD00064214 | |
| l-leucic acid, s-2-hydroxy-4-methylpentanoic acid, 2s-2-hydroxy-4-methylpentanoic acid, l-2-hydroxy-4-methylvaleric acid, s-leucic acid, s-2-hydroxy-4-methylvaleric acid, l-2-hydroxyisocaproic acid, l-alpha-hydroxyisocaproic acid, s---2-hydroxyisocaproic acid, pentanoic acid, 2-hydroxy-4-methyl-, 2s | |
| CC(C)CC(C(=O)O)O | |
| 132.159 | |
| CHEBI:44510 | |
| ≥99.0% (T) | |
| L-Leucic Acid |
| 81°C | |
| C6H12O3 | |
| 1 g | |
| LVRFTAZAXQPQHI-YFKPBYRVSA-N | |
| (2S)-2-hydroxy-4-methylpentanoic acid | |
| 83697 | |
| 132.16 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-1340
TSCA : Yes