missing translation for 'onlineSavingsMsg'
Learn More

L-Leucic Acid 99.0+%, TCI America™

Supplier:  TCI America L002625G

 View more versions of this product

Catalog No. L002625G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

L-Leucic Acid
81°C
C6H12O3
25 g
LVRFTAZAXQPQHI-YFKPBYRVSA-N
(2S)-2-hydroxy-4-methylpentanoic acid
83697
132.16
Crystalline Powder
13748-90-8
White
MFCD00064214
l-leucic acid, s-2-hydroxy-4-methylpentanoic acid, 2s-2-hydroxy-4-methylpentanoic acid, l-2-hydroxy-4-methylvaleric acid, s-leucic acid, s-2-hydroxy-4-methylvaleric acid, l-2-hydroxyisocaproic acid, l-alpha-hydroxyisocaproic acid, s---2-hydroxyisocaproic acid, pentanoic acid, 2-hydroxy-4-methyl-, 2s
CC(C)CC(C(=O)O)O
132.159
CHEBI:44510
≥99.0% (T)

Chemical Identifiers

13748-90-8
132.159
LVRFTAZAXQPQHI-YFKPBYRVSA-N
83697
(2S)-2-hydroxy-4-methylpentanoic acid
C6H12O3
MFCD00064214
l-leucic acid, s-2-hydroxy-4-methylpentanoic acid, 2s-2-hydroxy-4-methylpentanoic acid, l-2-hydroxy-4-methylvaleric acid, s-leucic acid, s-2-hydroxy-4-methylvaleric acid, l-2-hydroxyisocaproic acid, l-alpha-hydroxyisocaproic acid, s---2-hydroxyisocaproic acid, pentanoic acid, 2-hydroxy-4-methyl-, 2s
CHEBI:44510
CC(C)CC(C(=O)O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1340

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions