accessibility menu, dialog, popup

UserName

H3CCH3(S)OHOHO

CAS RN 13748-90-8

IUPAC Name: (2S)-2-hydroxy-4-methylpentanoic acid
Synonyms: (+)-α-hydroxyisocaproate L-2-hydroxyisocaproic acid hydroxyisocaproic acid (S)-2-hydroxyisocaproic acid L-α-hydroxyisocaproate L-2-hydroxyisocaproate (S)-leucic acid (+)-2-hydroxyisocaproic acid L-a-hydroxyisocaproic acid L-leucic acid
Molecular Weight (g/mol): 132.16
Molecular Formula: C6H12O3
InChi Key: LVRFTAZAXQPQHI-YFKPBYRVSA-N
SMILES: CC(C)C[C@H](O)C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)