missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-(-)-Lactide 98.0+%, TCI America™
Supplier: TCI America L0115250G
Specifications
| L-(-)-Lactide | |
| 4511-42-6 | |
| 250 g | |
| C6H8O4 | |
| l-lactide, 3s,6s-3,6-dimethyl-1,4-dioxane-2,5-dione, l---lactide, lactide, l, l-dilactide, 3s-cis-3,6-dimethyl-1,4-dioxane-2,5-dione, ll-lactide, unii-ij13to4no1, 1,4-dioxane-2,5-dione, 3,6-dimethyl-, 3s,6s, ij13to4no1 | |
| CC1OC(=O)C(C)OC1=O | |
| 144.13 | |
| 144.13 |
| 97°C | |
| White | |
| Crystal-Powder at 20°C | |
| MFCD00070594 | |
| JJTUDXZGHPGLLC-UHFFFAOYNA-N | |
| 3,6-dimethyl-1,4-dioxane-2,5-dione | |
| 107983 | |
| ≥98.0% (T) |
Chemical Identifiers
| 4511-42-6 | |
| 144.13 | |
| JJTUDXZGHPGLLC-UHFFFAOYNA-N | |
| 107983 | |
| CC1OC(=O)C(C)OC1=O |
| C6H8O4 | |
| MFCD00070594 | |
| l-lactide, 3s,6s-3,6-dimethyl-1,4-dioxane-2,5-dione, l---lactide, lactide, l, l-dilactide, 3s-cis-3,6-dimethyl-1,4-dioxane-2,5-dione, ll-lactide, unii-ij13to4no1, 1,4-dioxane-2,5-dione, 3,6-dimethyl-, 3s,6s, ij13to4no1 | |
| 3,6-dimethyl-1,4-dioxane-2,5-dione |
Safety and Handling
EINECSNumber : (5)-5295
TSCA : Yes