missing translation for 'onlineSavingsMsg'
Learn More

L-(-)-Lactide 98.0+%, TCI America™

Catalog No. p-7131826
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L011525G 25 g
L0115250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L011525G Supplier TCI America Supplier No. L011525G
Only null left

Chemical Identifiers

CAS 4511-42-6
Molecular Formula C6H8O4
Molecular Weight (g/mol) 144.13
MDL Number MFCD00070594
InChI Key JJTUDXZGHPGLLC-UHFFFAOYNA-N
Synonym l-lactide, 3s,6s-3,6-dimethyl-1,4-dioxane-2,5-dione, l---lactide, lactide, l, l-dilactide, 3s-cis-3,6-dimethyl-1,4-dioxane-2,5-dione, ll-lactide, unii-ij13to4no1, 1,4-dioxane-2,5-dione, 3,6-dimethyl-, 3s,6s, ij13to4no1
PubChem CID 107983
IUPAC Name 3,6-dimethyl-1,4-dioxane-2,5-dione
SMILES CC1OC(=O)C(C)OC1=O

Specifications

Melting Point 97°C
Color White
Quantity 25 g
Physical Form Crystal-Powder at 20°C
Formula Weight 144.13
Percent Purity ≥98.0% (T)
Chemical Name or Material L-(-)-Lactide
EINECSNumber (5)-5295
TSCA Yes