missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| L-Homoserine | |
| 194°C | |
| C4H9NO3 | |
| 5 g | |
| UKAUYVFTDYCKQA-VKHMYHEASA-N | |
| (2S)-2-amino-4-hydroxybutanoic acid | |
| 12647 | |
| 119.12 | |
| Crystalline Powder |
| 672-15-1 | |
| White | |
| MFCD00063090 | |
| l-homoserine, homoserine, s-2-amino-4-hydroxybutanoic acid, 2s-2-amino-4-hydroxybutanoic acid, h-hoser-oh, s-2-amino-4-hydroxybutyric acid, homoserine van, 2-amino-4-hydroxybutyric acid, s-homoserine, unii-6ka95x0ivo | |
| C(CO)C(C(=O)O)N | |
| 119.12 | |
| CHEBI:15699 | |
| ≥98.0% (T) |
Chemical Identifiers
| 672-15-1 | |
| 119.12 | |
| UKAUYVFTDYCKQA-VKHMYHEASA-N | |
| 12647 | |
| (2S)-2-amino-4-hydroxybutanoic acid |
| C4H9NO3 | |
| MFCD00063090 | |
| l-homoserine, homoserine, s-2-amino-4-hydroxybutanoic acid, 2s-2-amino-4-hydroxybutanoic acid, h-hoser-oh, s-2-amino-4-hydroxybutyric acid, homoserine van, 2-amino-4-hydroxybutyric acid, s-homoserine, unii-6ka95x0ivo | |
| CHEBI:15699 | |
| C(CO)C(C(=O)O)N |
Safety and Handling
EINECSNumber : (2)-2759
TSCA : No