missing translation for 'onlineSavingsMsg'
Learn More

L-Homoserine 98.0+%, TCI America™

Catalog No. p-7130872
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H10301G 1 g
H10305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H10301G Supplier TCI America Supplier No. H10301G
Only null left

Chemical Identifiers

CAS 672-15-1
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD00063090
InChI Key UKAUYVFTDYCKQA-VKHMYHEASA-N
Synonym l-homoserine, homoserine, s-2-amino-4-hydroxybutanoic acid, 2s-2-amino-4-hydroxybutanoic acid, h-hoser-oh, s-2-amino-4-hydroxybutyric acid, homoserine van, 2-amino-4-hydroxybutyric acid, s-homoserine, unii-6ka95x0ivo
PubChem CID 12647
ChEBI CHEBI:15699
IUPAC Name (2S)-2-amino-4-hydroxybutanoic acid
SMILES C(CO)C(C(=O)O)N

Specifications

Melting Point 194°C
Color White
Quantity 1 g
Formula Weight 119.12
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-Homoserine
EINECSNumber (2)-2759
TSCA No