missing translation for 'onlineSavingsMsg'
Learn More

L-Homoserine, 99%

CAS: 672-15-1 | C4H9NO3 | 119.12 g/mol

Supplier:  Thermo Scientific Chemicals L1242703

 View more versions of this product

Catalog No. AAL1242703


Only null left

Description

Description

L-Homoserine is used in the biosynthesis of methionine, threonine and isoleucine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Homoserine is used in the biosynthesis of methionine, threonine and isoleucine.

Solubility
Soluble in water (1100 mg/ml at 30°C), and methanol (Sparingly). Insoluble in ethanol.

Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents. Store at room temperature.
Specifications

Specifications

∽203°C (decomposition)
672-15-1
MFCD00063090
14,4739
Soluble in water (1100mg/ml at 30°C),and methanol (Sparingly). Insoluble in ethanol.
C(CO)C(C(=O)O)N
1 g
12647
−9° (c=2 in Water)
99%
L-Homoserine
C4H9NO3
1721681
l-homoserine, homoserine, s-2-amino-4-hydroxybutanoic acid, 2s-2-amino-4-hydroxybutanoic acid, h-hoser-oh, s-2-amino-4-hydroxybutyric acid, homoserine van, 2-amino-4-hydroxybutyric acid, s-homoserine, unii-6ka95x0ivo
UKAUYVFTDYCKQA-VKHMYHEASA-N
(2S)-2-amino-4-hydroxybutanoic acid
119.12
CHEBI:15699
119.12

Chemical Identifiers

672-15-1
119.12
UKAUYVFTDYCKQA-VKHMYHEASA-N
12647
(2S)-2-amino-4-hydroxybutanoic acid
C4H9NO3
MFCD00063090
l-homoserine, homoserine, s-2-amino-4-hydroxybutanoic acid, 2s-2-amino-4-hydroxybutanoic acid, h-hoser-oh, s-2-amino-4-hydroxybutyric acid, homoserine van, 2-amino-4-hydroxybutyric acid, s-homoserine, unii-6ka95x0ivo
CHEBI:15699
C(CO)C(C(=O)O)N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 211-590-6

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only