missing translation for 'onlineSavingsMsg'
Learn More

L-Homoserine, 99%

Catalog No. p-7045907
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL12427MD 250 mg
AAL1242703 1 g
AAL1242706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL12427MD Supplier Thermo Scientific Chemicals Supplier No. L12427MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 672-15-1 | C4H9NO3 | 119.12 g/mol

L-Homoserine is used in the biosynthesis of methionine, threonine and isoleucine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Homoserine is used in the biosynthesis of methionine, threonine and isoleucine.

Solubility
Soluble in water (1100 mg/ml at 30°C), and methanol (Sparingly). Insoluble in ethanol.

Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents. Store at room temperature.

Chemical Identifiers

CAS 672-15-1
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD00063090
InChI Key UKAUYVFTDYCKQA-VKHMYHEASA-N
Synonym l-homoserine, homoserine, s-2-amino-4-hydroxybutanoic acid, 2s-2-amino-4-hydroxybutanoic acid, h-hoser-oh, s-2-amino-4-hydroxybutyric acid, homoserine van, 2-amino-4-hydroxybutyric acid, s-homoserine, unii-6ka95x0ivo
PubChem CID 12647
ChEBI CHEBI:15699
IUPAC Name (2S)-2-amino-4-hydroxybutanoic acid
SMILES C(CO)C(C(=O)O)N

Specifications

Melting Point ∽203°C (decomposition)
Beilstein 1721681
Merck Index 14,4739
Solubility Information Soluble in water (1100mg/ml at 30°C),and methanol (Sparingly). Insoluble in ethanol.
Quantity 250 mg
Formula Weight 119.12
Optical Rotation −9° (c=2 in Water)
Percent Purity 99%
Chemical Name or Material L-Homoserine
EINECSNumber 211-590-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only