Learn More
Thermo Scientific Chemicals L-Histidine monohydrochloride monohydrate, 99%
L-Histidine monohydrochloride monohydrate, CAS # 5934-29-2, is a histidine precursor through histidine decarboxylase. | CAS: 5934-29-2 | C6H12ClN3O3 | 209.63 g/mol
Supplier: Thermo Scientific Chemicals A1762730
Description
L-Histidine monohydrochloride monohydrate is used as a precursor through histidine decarboxylase. It is also used to study the cultures of the human T-lymphoblastic leukemia cell line, modulation of apoptosis. It is also used to enhance the biosynthesis of Lovastatin by cultured aspergillus terreus. It is utilized as a single nitrogen source to probe swarming in Pseudomonas aeruginosa on agar. It is employed as a component to avoid the risk of contaminating bioproducts with adventitious viruses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• L-Histidine monohydrochloride monohydrate is a histidine precursor via histidine decarboxylase.
Application
• As a precursor of histidine, it can be used to enhance the biosyntyhesis of lovastatin by cultured Aspergillus terreus
• It is utilized as a single nitrogen source to probe swarming in Pseudomonas aeruginosa on agar
• It can be employed in laboraotry applications as a component to avoid the risk of contaminating bioproducts with adventitious viruses
• It is also used for laboratory study of cultures of the human T-lymphoblastic leukemia cell line, and modulation of apoptosis
Specifications
| ∽240°C (decomposition) | |
| 5934-29-2 | |
| C6H12ClN3O3 | |
| 4168261 | |
| l-histidine hydrochloride hydrate, l-histidine hydrochloride monohydrate, s-2-amino-3-1h-imidazol-4-yl propanoic acid hydrochloride hydrate, h-his-oh.hcl.h2o, l-histidine monohydrochloride monohydrate, l-histidine, monohydrochloride, monohydrate, l-histidin hydrate hydrochloride, histidine, monohydrochloride, monohydrate, l, h-his-ohhclh2o, l-histidine, hydrochloride, monohydrate | |
| CMXXUDSWGMGYLZ-XRIGFGBMSA-N | |
| (2S)-2-amino-3-(1H-imidazol-4-yl)propanoic acid hydrate hydrochloride | |
| 209.63 | |
| 209.63 (191.62 Anhydrous) |
| L-Histidine Monohydrochloride Monohydrate | |
| 99% | |
| MFCD00151027 | |
| 14,4720 | |
| Soluble in water. | |
| O.Cl.N[C@@H](CC1=CNC=N1)C(O)=O | |
| 250 g | |
| 165377 | |
| 99% |
Chemical Identifiers
| 5934-29-2 | |
| 209.63 | |
| CMXXUDSWGMGYLZ-XRIGFGBMSA-N | |
| 165377 | |
| O.Cl.N[C@@H](CC1=CNC=N1)C(O)=O |
| C6H12ClN3O3 | |
| MFCD00151027 | |
| l-histidine hydrochloride hydrate, l-histidine hydrochloride monohydrate, s-2-amino-3-1h-imidazol-4-yl propanoic acid hydrochloride hydrate, h-his-oh.hcl.h2o, l-histidine monohydrochloride monohydrate, l-histidine, monohydrochloride, monohydrate, l-histidin hydrate hydrochloride, histidine, monohydrochloride, monohydrate, l, h-his-ohhclh2o, l-histidine, hydrochloride, monohydrate | |
| (2S)-2-amino-3-(1H-imidazol-4-yl)propanoic acid hydrate hydrochloride |
Safety and Handling
EINECSNumber : 211-438-9
RTECSNumber : MS3119000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only