missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Histidine monohydrochloride monohydrate, 99%

Catalog No. p-3345395
Change view
Click to view available options
Quantity:
50 g
250 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1762718 50 g
AAA1762730 250 g
AAA1762736 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1762718 Supplier Thermo Scientific Chemicals Supplier No. A1762718

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

L-Histidine monohydrochloride monohydrate, CAS # 5934-29-2, is a histidine precursor through histidine decarboxylase. | CAS: 5934-29-2 | C6H12ClN3O3 | 209.63 g/mol

L-Histidine monohydrochloride monohydrate is used as a precursor through histidine decarboxylase. It is also used to study the cultures of the human T-lymphoblastic leukemia cell line, modulation of apoptosis. It is also used to enhance the biosynthesis of Lovastatin by cultured aspergillus terreus. It is utilized as a single nitrogen source to probe swarming in Pseudomonas aeruginosa on agar. It is employed as a component to avoid the risk of contaminating bioproducts with adventitious viruses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• L-Histidine monohydrochloride monohydrate is a histidine precursor via histidine decarboxylase.

Application

• As a precursor of histidine, it can be used to enhance the biosyntyhesis of lovastatin by cultured Aspergillus terreus
• It is utilized as a single nitrogen source to probe swarming in Pseudomonas aeruginosa on agar
• It can be employed in laboraotry applications as a component to avoid the risk of contaminating bioproducts with adventitious viruses
• It is also used for laboratory study of cultures of the human T-lymphoblastic leukemia cell line, and modulation of apoptosis

Chemical Identifiers

CAS 5934-29-2
Molecular Formula C6H12ClN3O3
Molecular Weight (g/mol) 209.63
MDL Number MFCD00151027
InChI Key CMXXUDSWGMGYLZ-XRIGFGBMSA-N
Synonym l-histidine hydrochloride hydrate, l-histidine hydrochloride monohydrate, s-2-amino-3-1h-imidazol-4-yl propanoic acid hydrochloride hydrate, h-his-oh.hcl.h2o, l-histidine monohydrochloride monohydrate, l-histidine, monohydrochloride, monohydrate, l-histidin hydrate hydrochloride, histidine, monohydrochloride, monohydrate, l, h-his-ohhclh2o, l-histidine, hydrochloride, monohydrate
PubChem CID 165377
IUPAC Name (2S)-2-amino-3-(1H-imidazol-4-yl)propanoic acid hydrate hydrochloride
SMILES O.Cl.N[C@@H](CC1=CNC=N1)C(O)=O

Specifications

Melting Point ∽240°C (decomposition)
Assay Percent Range 99%
Beilstein 4168261
Merck Index 14,4720
Solubility Information Soluble in water.
Quantity 50 g
Formula Weight 209.63 (191.62 anhydrous)
Percent Purity 99%
Chemical Name or Material L-Histidine Monohydrochloride Monohydrate
EINECSNumber 211-438-9
RTECSNumber MS3119000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only