missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Histidine, free base, ≥99%, MP Biomedicals™
Precursor of histamine by action of histidine decarboxylase
Manufacturer: Mp Biomedicals 0210195490
Specifications
L-Histidine | |
≥99% | |
MFCD00064315 | |
HNDVDQJCIGZPNO-YFKPBYRVSA-N | |
(2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid | |
6274 | |
155.16 | |
White |
71-00-1 | |
C6H9N3O2 | |
anti-rheuma, glyoxaline-5-alanine, h-his-oh, histidine, istidina, l---histidine, l-histidin, l-histidine, s-histidine | |
C1=C(NC=N1)CC(C(=O)O)N | |
155.157 | |
CHEBI:15971 | |
Crystalline Powder | |
500g |
Chemical Identifiers
71-00-1 | |
155.157 | |
HNDVDQJCIGZPNO-YFKPBYRVSA-N | |
6274 | |
(2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid |
C6H9N3O2 | |
MFCD00064315 | |
anti-rheuma, glyoxaline-5-alanine, h-his-oh, histidine, istidina, l---histidine, l-histidin, l-histidine, s-histidine | |
CHEBI:15971 | |
C1=C(NC=N1)CC(C(=O)O)N |
Safety and Handling
EINECSNumber : 200-745-3
Recommended Storage : Store at Room Temperature.