accessibility menu, dialog, popup

UserName

H2NNHNHOO

CAS RN 71-00-1

IUPAC Name: 2-amino-3-(1H-imidazol-5-yl)propanoic acid
Synonyms: histidine
Molecular Weight (g/mol): 155.16
Molecular Formula: C6H9N3O2
InChi Key: HNDVDQJCIGZPNO-UHFFFAOYNA-N
SMILES: NC(CC1=CN=CN1)C(O)=O
Molecular Weight (g/mol) 
  • (4)
  • (15)
Quantity 
  • (3)
  • (6)
  • (5)
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (11)
Grade 
  • (1)
  • (4)
  • (4)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (17)
  • (11)
  • (1)
  • (2)
  • (11)
  • (6)
  • (4)
  • (15)
  • (3)
  • (6)
  • (5)
  • (2)
  • (3)
  • (3)
  • (3)
  • (11)
  • (1)
  • (4)
  • (4)
  • (3)