missing translation for 'onlineSavingsMsg'
Learn More

L-Histidine, free base, ≥99%, MP Biomedicals™

Catalog No. p-4404399
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10195480 100 g
ICN10195490 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10195480 Supplier MP Biomedicals Inc Supplier No. 0210195480
Only null left

Precursor of histamine by action of histidine decarboxylase

Chemical Identifiers

CAS 71-00-1
Molecular Formula C6H9N3O2
Molecular Weight (g/mol) 155.16
MDL Number MFCD00064315
InChI Key HNDVDQJCIGZPNO-UHFFFAOYNA-N
Synonym l-histidine, histidine, h-his-oh, glyoxaline-5-alanine, anti-rheuma, l---histidine, istidina, s-histidine, l-histidin
PubChem CID 6274
ChEBI CHEBI:15971
SMILES NC(CC1=CN=CN1)C(O)=O

Specifications

Color White
Assay Percent Range ≥99%
IUPAC Name 2-amino-3-(1H-imidazol-5-yl)propanoic acid
Quantity 100 g
Formula Weight 155.16
Physical Form Crystalline Powder
Chemical Name or Material L-Histidine
EINECSNumber 200-745-3
Recommended Storage Store at Room Temperature.