missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Histidine, 98+%

CAS: 71-00-1 | C6H9N3O2 | 155.16 g/mol

$67.55 - $626.74

Chemical Identifiers

CAS 71-00-1
Molecular Formula C6H9N3O2
Molecular Weight (g/mol) 155.16
MDL Number MFCD00064315
InChI Key HNDVDQJCIGZPNO-UHFFFAOYNA-N
Synonym l-histidine, histidine, h-his-oh, glyoxaline-5-alanine, anti-rheuma, l---histidine, istidina, s-histidine, l-histidin
PubChem CID 6274
ChEBI CHEBI:15971
SMILES NC(CC1=CN=CN1)C(O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAA1041314
View Documents Promotion Details
Thermo Scientific Chemicals
A1041314
25 g
Each for $67.55
Only null left
 
AAA1041322
View Documents Promotion Details
Thermo Scientific Chemicals
A1041322
100 g
Each for $174.10
Only null left
 
AAA1041336
View Documents Promotion Details
Thermo Scientific Chemicals
A1041336
500 g
Each for $626.74
Only null left
 
Description

Description

L-Histidine acts as a precursor to histamine and a component of carnosine. It is also used in medicine, feed additive, biochemical research, dietary supplement. It is used in a nutrition enhancer, is the amino acid fluid infusion. It can be used in the treatment of gastric ulcer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Histidine acts as a precursor to histamine and a component of carnosine. It is also used in medicine, feed additive, biochemical research, dietary supplement. It is used in a nutrition enhancer, is the amino acid fluid infusion. It can be used in the treatment of gastric ulcer.

Solubility
Soluble in water (15 mg/ml at 20°C).

Notes
Store at 4°C.Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

71-00-1
155.16
HNDVDQJCIGZPNO-UHFFFAOYNA-N
6274
NC(CC1=CN=CN1)C(O)=O
C6H9N3O2
MFCD00064315
l-histidine, histidine, h-his-oh, glyoxaline-5-alanine, anti-rheuma, l---histidine, istidina, s-histidine, l-histidin
CHEBI:15971

Specifications

∽281°C (decomposition)
>98%
MFCD00064315
14,4720
Soluble in water (15mg/ml at 20°C).
NC(CC1=CN=CN1)C(O)=O
155.16
CHEBI:15971
≥98%
71-00-1
C6H9N3O2
4673585
l-histidine, histidine, h-his-oh, glyoxaline-5-alanine, anti-rheuma, l---histidine, istidina, s-histidine, l-histidin
HNDVDQJCIGZPNO-UHFFFAOYNA-N
(2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid
6274
155.16
L-Histidine
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-745-3

RTECSNumber : MS3070000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only