missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals L-Histidine, 98+%
Description
L-Histidine acts as a precursor to histamine and a component of carnosine. It is also used in medicine, feed additive, biochemical research, dietary supplement. It is used in a nutrition enhancer, is the amino acid fluid infusion. It can be used in the treatment of gastric ulcer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
L-Histidine acts as a precursor to histamine and a component of carnosine. It is also used in medicine, feed additive, biochemical research, dietary supplement. It is used in a nutrition enhancer, is the amino acid fluid infusion. It can be used in the treatment of gastric ulcer.
Solubility
Soluble in water (15 mg/ml at 20°C).
Notes
Store at 4°C.Incompatible with oxidizing agents.
L-Histidine acts as a precursor to histamine and a component of carnosine. It is also used in medicine, feed additive, biochemical research, dietary supplement. It is used in a nutrition enhancer, is the amino acid fluid infusion. It can be used in the treatment of gastric ulcer.
Solubility
Soluble in water (15 mg/ml at 20°C).
Notes
Store at 4°C.Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | ∼281°C (decomposition) |
| pH | 7.0 to 8 |
| Assay Percent Range | >98% |
| Beilstein | 4673585 |
| Merck Index | 14,4720 |
| Solubility Information | Soluble in water (15mg/ml at 20°C). |
| IUPAC Name | 2-amino-3-(1H-imidazol-5-yl)propanoic acid |
| Quantity | 100 g |
| Odor | Odorless |
| Formula Weight | 155.16 |
| Show More |
Safety and Handling
| EINECSNumber | 200-745-3 |
| RTECSNumber | MS3070000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only