missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Histidine, 98+%

CAS: 71-00-1 | C6H9N3O2 | 155.16 g/mol

Supplier:  Thermo Scientific Chemicals A1041336

 View more versions of this product

Catalog No. AAA1041336


Only null left

Description

Description

L-Histidine acts as a precursor to histamine and a component of carnosine. It is also used in medicine, feed additive, biochemical research, dietary supplement. It is used in a nutrition enhancer, is the amino acid fluid infusion. It can be used in the treatment of gastric ulcer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Histidine acts as a precursor to histamine and a component of carnosine. It is also used in medicine, feed additive, biochemical research, dietary supplement. It is used in a nutrition enhancer, is the amino acid fluid infusion. It can be used in the treatment of gastric ulcer.

Solubility
Soluble in water (15 mg/ml at 20°C).

Notes
Store at 4°C.Incompatible with oxidizing agents.
Specifications

Specifications

∼281°C (decomposition)
71-00-1
>98%
MFCD00064315
14,4720
Soluble in water (15mg/ml at 20°C).
NC(CC1=CN=CN1)C(O)=O
155.16
6274
Odorless
155.16
L-Histidine
7.0 to 8
C6H9N3O2
4673585
l-histidine, histidine, h-his-oh, glyoxaline-5-alanine, anti-rheuma, l---histidine, istidina, s-histidine, l-histidin
HNDVDQJCIGZPNO-UHFFFAOYNA-N
(2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid
500 g
CHEBI:15971
−39° (c=5 in water)
≥98%

Chemical Identifiers

71-00-1
155.16
HNDVDQJCIGZPNO-UHFFFAOYNA-N
6274
NC(CC1=CN=CN1)C(O)=O
C6H9N3O2
MFCD00064315
l-histidine, histidine, h-his-oh, glyoxaline-5-alanine, anti-rheuma, l---histidine, istidina, s-histidine, l-histidin
CHEBI:15971
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-745-3

RTECSNumber : MS3070000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only