missing translation for 'onlineSavingsMsg'
Learn More

L-Histidine, 98.5-101.5%, Spectrum™ Chemical

$210.75 - $12,154.39

Chemical Identifiers

CAS 71-00-1
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
18611283
View Documents Promotion Details
N/A 25 g Poly Pail
Each for $210.75
Only null left
 
18611282
View Documents Promotion Details
N/A 100 g Poly Pail
Each for $583.86
Only null left
 
18611281
View Documents Promotion Details
N/A 1 kg Poly Pail
Each for $3,491.04
Only null left
 
18611284
View Documents Promotion Details
N/A 5 kg Poly Pail N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

71-00-1

Specifications

71-00-1
1.5e-05
+12.6° to +14.0°
Reagent
1
0.002
98.5 to 101.5%
L-Histidine
Videos
SDS
Documents

Documents

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.